Assay Details
Target Gene Details
Entrez Gene ID: | 20630 |
Gene Name: | U1 small nuclear ribonucleoprotein C |
Gene Aliases: |
Snrp1c, U1-C, U1C |
Location: |
Chr.17:27840087-27851968 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snrpc | NM_011432.2 | NP_035562.1 | ||
| AK137213.1 | BAE23273.1 | |||
| AK168973.1 | BAE40776.1 | |||
| BC008243.1 | AAH08243.1 | |||
| BC092266.1 | AAH92266.1 | |||
| CJ085691.1 | ||||
| U70315.1 | AAB08894.1 | |||
| X96767.1 | CAA65542.1 |

Genomic Map