Assay Details
Target Gene Details
Entrez Gene ID: | 21678 |
Gene Name: | TEA domain family member 3 |
Gene Aliases: |
DTEF-1, ETFR-1, TEAD-3, TEF-5, Tcf13r2 |
Location: |
Chr.17:28331671-28350805 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tead3 | NM_011566.4 | 1 | 55 | NP_035696.3 |
| AK031555.1 | BAC27448.1 | |||
| CF617964.1 | 1 | 55 | ||
| D87964.1 | BAA13517.1 |
More Information
Set Membership: |
|

Genomic Map