Assay Details
Target Gene Details
Entrez Gene ID: | 66972 |
Gene Name: | solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23 |
Gene Aliases: |
2310067G05Rik, SCaMC-3 |
Location: |
Chr.17:57043711-57059880 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a23 | NM_025877.4 | NP_080153.2 | ||
| AK158821.1 | 2 | 1776 | ||
| AK199511.1 | ||||
| AK206310.1 | ||||
| BC072660.1 | AAH72660.1 |
More Information
Set Membership: |
|

Genomic Map