Assay Details
Target Gene Details
Entrez Gene ID: | 21678 |
Gene Name: | TEA domain family member 3 |
Gene Aliases: |
DTEF-1, ETFR-1, TEAD-3, TEF-5, Tcf13r2 |
Location: |
Chr.17:28331671-28350805 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tead3 | NM_001098226.3 | 11 | 1064 | NP_001091696.2 |
| NM_001204156.1 | 11 | 1387 | NP_001191085.1 | |
| NM_011566.4 | 11 | 1228 | NP_035696.3 | |
| AF002670.1 | 11 | 1183 | AAC12263.1 | |
| AK087649.1 | 11 | 1066 | ||
| AK143551.1 | 10 | 2100 | ||
| BC021364.1 | 11 | 1108 | AAH21364.1 | |
| BC027383.1 | 2 | 139 | AAH27383.1 | |
| BC039984.1 | 2 | 174 | AAH39984.1 | |
| D87963.1 | 10 | 898 | BAA13516.1 | |
| D87964.1 | 11 | 1209 | BAA13517.1 | |
| Y10027.1 | 10 | 990 | CAA71136.1 | |
| Y16611.1 | 10 | 981 | CAA76315.2 |
More Information
Set Membership: |
|

Genomic Map