Assay Details
Target Gene Details
Entrez Gene ID: | 60532 |
Gene Name: | Wilms tumour 1-associating protein |
Gene Aliases: |
2810408K05Rik, 9430038B09Rik |
Location: |
Chr.17:12966798-12992808 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wtap | NM_001113532.1 | NP_001107004.1 | ||
| NM_001113533.1 | NP_001107005.1 | |||
| NM_175394.2 | NP_780603.1 | |||
| AJ276707.1 | CAC16790.1 | |||
| AK031965.1 | ||||
| AK076111.1 | BAC36191.1 | |||
| AK156334.1 | BAE33680.1 | |||
| BC046416.1 | AAH46416.1 | |||
| BC093504.1 | AAH93504.1 | |||
| BY016042.1 |
More Information
Set Membership: |
|

Genomic Map