Assay Details
Target Gene Details
Entrez Gene ID: | 22436 |
Gene Name: | xanthine dehydrogenase |
Gene Aliases: |
XO, Xor, Xox-1, Xox1 |
Location: |
Chr.17:73883895-73950196 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xdh | NM_011723.3 | NP_035853.2 | ||
| AK034256.1 | ||||
| AK090058.1 | ||||
| AK134156.1 | ||||
| AK143412.1 | ||||
| AK144700.1 | BAE26022.1 | |||
| AK160010.1 | ||||
| AK161529.1 | ||||
| BC141183.1 | ||||
| X62932.1 | CAA44705.1 |
More Information
Set Membership: |
|

Genomic Map