Assay Details
Target Gene Details
Entrez Gene ID: | 20656 |
Gene Name: | superoxide dismutase 2, mitochondrial |
Gene Aliases: |
MnSOD, Sod-2 |
Location: |
Chr.17:13007839-13018119 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sod2 | NM_013671.3 | 1 | 159 | NP_038699.2 |
| AK002428.1 | BAB22095.1 | |||
| AK002534.1 | BAB22170.1 | |||
| AK012354.1 | BAB28183.1 | |||
| AK135435.1 | 1 | 148 | BAE22531.1 | |
| AK152047.1 | BAE30903.1 | |||
| AK167521.1 | BAE39593.1 | |||
| AK168082.1 | BAE40055.1 | |||
| AK168591.1 | 1 | 151 | BAE40460.1 | |
| BC010548.1 | AAH10548.1 | |||
| BY137511.1 | 1 | 159 | ||
| X04972.1 | CAA28645.1 | |||
| Z18857.1 | CAA79308.1 |
More Information
Set Membership: |
|

Genomic Map