Assay Details
Target Gene Details
Entrez Gene ID: | 224481 |
Gene Name: | transcription factor B1, mitochondrial |
Gene Aliases: |
AI429207 |
Location: |
Chr.17:3519256-3557822 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tfb1m | NM_146074.1 | NP_666186.1 | ||
| AF508971.1 | AAM33651.1 | |||
| BC032930.1 | AAH32930.1 |
More Information
Set Membership: |
|

Genomic Map