Assay Details
Target Gene Details
Entrez Gene ID: | 224661 |
Gene Name: | solute carrier family 26, member 8 |
Gene Aliases: |
- |
Location: |
Chr.17:28637779-28691237 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a8 | NM_001290320.1 | NP_001277249.1 | ||
| BC027076.1 | AAH27076.1 |
More Information
Set Membership: |
|

Genomic Map