Assay Details
Target Gene Details
Entrez Gene ID: | 381112 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 33 |
Gene Aliases: |
Gm941 |
Location: |
Chr.17:80306930-80427079 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef33 |
Target Gene Details
Entrez Gene ID: | 20662 |
Gene Name: | son of sevenless homolog 1 (Drosophila) |
Gene Aliases: |
4430401P03Rik, 9630010N06, AI449023 |
Location: |
Chr.17:80393752-80480453 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sos1 | NM_009231.2 | NP_033257.2 | ||
| AK140310.1 | BAE24327.1 |
More Information
Set Membership: |
|

Genomic Map