Assay Details
Target Gene Details
Entrez Gene ID: | 22436 |
Gene Name: | xanthine dehydrogenase |
Gene Aliases: |
XO, Xor, Xox-1, Xox1 |
Location: |
Chr.17:73883895-73950196 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xdh | NM_011723.3 | 32 | 3546 | NP_035853.2 |
| AK008517.1 | 6 | 463 | BAB25715.1 | |
| AK034256.1 | 32 | 3494 | ||
| AK036869.1 | 7 | 526 | ||
| AK090058.1 | 23 | 2688 | ||
| AK134156.1 | 18 | 2031 | ||
| AK143412.1 | 32 | 3495 | ||
| AK144700.1 | 32 | 3498 | BAE26022.1 | |
| AK160010.1 | 29 | 3245 | ||
| AK161529.1 | 32 | 3497 | ||
| BC003997.1 | AAH03997.1 | |||
| BC141183.1 | 32 | 3521 | ||
| X62932.1 | 32 | 3480 | CAA44705.1 |
More Information
Set Membership: |
|

Genomic Map