Assay Details
Target Gene Details
Entrez Gene ID: | 22436 |
Gene Name: | xanthine dehydrogenase |
Gene Aliases: |
XO, Xor, Xox-1, Xox1 |
Location: |
Chr.17:73883895-73950196 on Build GRCm38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xdh | NM_011723.3 | 34 | 3792 | NP_035853.2 |
| AK008517.1 | 8 | 709 | BAB25715.1 | |
| AK034256.1 | 34 | 3740 | ||
| AK036869.1 | 9 | 772 | ||
| AK090058.1 | 25 | 2934 | ||
| AK134156.1 | 20 | 2277 | ||
| AK143412.1 | 34 | 3741 | ||
| AK144700.1 | 34 | 3744 | BAE26022.1 | |
| AK160010.1 | 31 | 3491 | ||
| AK161529.1 | 34 | 3743 | ||
| BC003997.1 | 3 | 264 | AAH03997.1 | |
| BC141183.1 | 34 | 3767 | ||
| X62932.1 | 34 | 3726 | CAA44705.1 |
More Information
Set Membership: |
|

Genomic Map