Assay Details
Target Gene Details
Entrez Gene ID: | 11789 |
Gene Name: | adenomatosis polyposis coli |
Gene Aliases: |
AI047805, AU020952, AW124434, CC1, Min, mAPC |
Location: |
Chr.18:34220307-34322190 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apc | NM_007462.3 | 4 | 620 | NP_031488.2 |
AK042021.1 | 4 | 621 | BAC31134.1 | |
AK045053.1 | 4 | 591 | BAC32198.1 | |
AK046280.1 | 4 | 611 | ||
AK080907.1 | 4 | 620 | BAC38073.1 | |
AK082724.1 | 4 | 366 | BAC38587.1 | |
BC141141.1 | 3 | 333 | ||
M88127.1 | 3 | 309 | AAB59632.1 |
