Assay Details
Target Gene Details
Entrez Gene ID: | 56386 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 6 |
Gene Aliases: |
AA536803, AU022389 |
Location: |
Chr.18:20684599-20746404 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galt6 | NM_019737.2 | NP_062711.1 | ||
| AF097158.1 | AAD41694.1 | |||
| AF142674.1 | AAF22224.1 | |||
| AK137385.1 | BAE23337.1 | |||
| AK138247.1 | BAE23594.1 | |||
| AK138610.1 | BAE23718.1 | |||
| AK144997.1 | BAE26177.1 | |||
| AK163956.1 | BAE37551.1 | |||
| AK164262.1 | BAE37708.1 | |||
| AK164724.1 | ||||
| BC011149.1 | AAH11149.1 |
More Information
Set Membership: |
|

Genomic Map