Assay Details
Target Gene Details
Entrez Gene ID: | 319888 |
Gene Name: | O-acyltransferase like |
Gene Aliases: |
5330437I02Rik, Acyl3 |
Location: |
Chr.18:65697370-65751624 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oacyl | NM_177028.3 | 4 | 495 | NP_796002.1 |
| AK030604.1 | 4 | 495 | BAC27041.1 | |
| AK030615.1 | 2 | 275 | BAC27048.1 | |
| BC141265.1 | 4 | 492 |
More Information
Set Membership: |
|

Genomic Map