Assay Details
Target Gene Details
Entrez Gene ID: | 30948 |
Gene Name: | bridging integrator 1 |
Gene Aliases: |
ALP-1, Amphl, BRAMP-2, SH3P9 |
Location: |
Chr.18:32377217-32435740 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bin1 | NM_001083334.1 | 6 | 660 | NP_001076803.1 |
NM_009668.2 | 6 | 660 | NP_033798.1 | |
BC065160.1 | 6 | 632 | AAH65160.1 | |
BQ573238.1 | ||||
BU707946.1 | 5 | 362 | ||
U60884.1 | 6 | 648 | AAC52661.1 |
