Assay Details
Target Gene Details
Entrez Gene ID: | 13664 |
Gene Name: | eukaryotic translation initiation factor 1A |
Gene Aliases: |
C76390, Ef1a, Eftu, Eif4c, eIF-1A, eIF-4C |
Location: |
Chr.18:46597658-46610225 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif1a | NM_010120.5 | NP_034250.3 | ||
| AF026481.1 | AAC63934.1 | |||
| AK009026.1 | BAB26034.1 | |||
| AK013268.1 | BAB28759.1 | |||
| AK019451.1 | BAB31727.1 | |||
| AK150651.1 | BAE29738.1 | |||
| AK150673.1 | BAE29755.1 | |||
| AK150677.1 | BAE29757.1 | |||
| AK167333.1 | BAE39435.1 | |||
| BC027437.1 | AAH27437.1 |
More Information
Set Membership: |
|

Genomic Map