Assay Details
Target Gene Details
Entrez Gene ID: | 328971 |
Gene Name: | serine peptidase inhibitor, Kazal type 10 |
Gene Aliases: |
A230091H23Rik |
Location: |
Chr.18:62548911-62662933 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spink10 | NM_177829.3 | 5 | 401 | NP_808497.1 |
| AK039062.1 | 5 | 402 | BAC30226.1 | |
| AK162840.1 | 5 | 369 | ||
| BC125360.1 | 5 | 317 | ||
| BC125362.1 | 5 | 317 | ||
| DQ437330.1 | 5 | 401 |
Target Gene Details
Entrez Gene ID: | 100038417 |
Gene Name: | serine peptidase inhibitor, Kazal type 13 |
Gene Aliases: |
ENSMUSG00000073551, Gm10534, Spink5l3 |
Location: |
Chr.18:62605898-62741387 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spink13 | NM_001168423.2 | NP_001161895.1 | ||
| AK139149.1 | BAE23902.1 |
More Information
Set Membership: |
|

Genomic Map