Assay Details
Target Gene Details
Entrez Gene ID: | 22240 |
Gene Name: | dihydropyrimidinase-like 3 |
Gene Aliases: |
CRMP-4, DRP-3, TUC4, ULIP-1, Ulip, Ulip1 |
Location: |
Chr.18:43320979-43438286 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpysl3 | NM_001136086.2 | 11 | 1415 | NP_001129558.1 |
| NM_001291455.1 | 11 | 1665 | NP_001278384.1 | |
| NM_009468.5 | 11 | 1596 | NP_033494.1 | |
| AK077000.1 | 11 | 1406 | ||
| AK133040.1 | 11 | 1415 | BAE21480.1 | |
| AK149024.1 | 11 | 1433 | ||
| AK161509.1 | 11 | 1337 | BAE36433.1 | |
| AK171653.1 | 4 | 496 | BAE42590.1 | |
| BC023003.1 | 11 | 1280 | AAH23003.1 | |
| X87817.1 | 11 | 1556 | CAA61082.1 |
More Information
Set Membership: |
|

Genomic Map