Assay Details
Target Gene Details
Entrez Gene ID: | 58523 |
Gene Name: | elongator acetyltransferase complex subunit 2 |
Gene Aliases: |
AU023723, Epl2, StIP1, Statip1 |
Location: |
Chr.18:24603923-24638830 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elp2 | NM_021448.2 | NP_067423.2 | ||
| AF291064.1 | AAG01032.1 | |||
| AK033246.1 | BAC28210.1 | |||
| AK034457.1 | BAC28716.1 | |||
| AK050960.1 | BAC34477.1 | |||
| AK167725.1 | BAE39766.1 | |||
| AK167853.1 | BAE39871.1 | |||
| AK168370.1 | BAE40302.1 | |||
| BC015284.1 | AAH15284.1 | |||
| BC051943.1 | AAH51943.1 |

Genomic Map