Assay Details
Target Gene Details
Entrez Gene ID: | 15562 |
Gene Name: | 5 hydroxytryptamine (serotonin) receptor 4 |
Gene Aliases: |
5-HT-4, 5-HT4, 5-HT<4L>, 5HTR4 |
Location: |
Chr.18:62322160-62498026 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htr4 | NM_008313.4 | NP_032339.2 | ||
| AJ011369.1 | CAA09598.1 | |||
| AK141587.1 | BAE24750.1 | |||
| AK141625.1 | BAE24773.1 | |||
| JN418927.1 | ||||
| JN418928.1 | ||||
| JN418929.1 | ||||
| JN418930.1 | ||||
| Y09585.1 | CAA70773.1 | |||
| Y09587.1 | CAA70775.1 | |||
| Y09588.1 | CAA70776.1 |
More Information
Set Membership: |
|

Genomic Map