Assay Details
Target Gene Details
Entrez Gene ID: | 72930 |
Gene Name: | protein phosphatase 2, regulatory subunit B, beta |
Gene Aliases: |
2900026H06Rik, 6330404L05Rik, E130009M08Rik, PP2A-PR55B, PR55-BETA, SCA12 |
Location: |
Chr.18:42645221-43059471 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r2b | NM_028392.3 | 3 | 498 | NP_082668.1 |
| NR_073583.1 | 3 | 950 | ||
| AF512670.1 | 3 | 204 | AAM46987.1 | |
| AF536771.1 | 3 | 267 | AAN05641.1 | |
| AK013600.1 | 3 | 500 | BAB28921.1 | |
| AK018119.1 | 3 | 455 | BAB31079.1 | |
| AK039592.1 | 3 | 341 | BAC30395.1 | |
| AK086561.1 | 1 | 687 | ||
| AK148956.1 | 3 | 469 | BAE28701.1 | |
| BC026686.1 | 3 | 436 | AAH26686.1 | |
| BC088979.1 | 3 | 433 | AAH88979.1 |
More Information
Set Membership: |
|

Genomic Map