Assay Details
Target Gene Details
Entrez Gene ID: | 20496 |
Gene Name: | solute carrier family 12, member 2 |
Gene Aliases: |
9330166H04Rik, Nkcc1, mBSC2, sy-ns |
Location: |
Chr.18:57878513-57946821 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc12a2 | NM_009194.3 | 22 | 3143 | NP_033220.2 |
| AK134606.1 | 6 | 571 | ||
| CN711123.1 | ||||
| U13174.1 | 22 | 3140 | AAC77832.1 |
More Information
Set Membership: |
|

Genomic Map