Assay Details
Target Gene Details
Entrez Gene ID: | 72432 |
Gene Name: | serine peptidase inhibitor, Kazal type 5 |
Gene Aliases: |
2310065D10Rik, AV238912, LEKT1, LETK1, VAKT1 |
Location: |
Chr.18:43959545-44022487 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spink5 | NM_001081180.1 | NP_001074649.1 | ||
| AJ426449.2 | CAD20045.2 | |||
| AK028727.1 | ||||
| AK035690.1 | BAC29153.1 | |||
| BC118011.1 |
More Information
Set Membership: |
|

Genomic Map