Assay Details
Target Gene Details
Entrez Gene ID: | 240334 |
Gene Name: | prenylcysteine oxidase 1 like |
Gene Aliases: |
BC060677, C630049M13 |
Location: |
Chr.18:61696164-61707635 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcyox1l | NM_172832.4 | 4 | 629 | NP_766420.1 |
| AK050006.1 | 4 | 630 | BAC34029.1 | |
| BC060677.1 | 4 | 619 | AAH60677.1 |
More Information
Set Membership: |
|

Genomic Map