Assay Details
Target Gene Details
Entrez Gene ID: | 27528 |
Gene Name: | neuronal regeneration related protein |
Gene Aliases: |
AI325076, D0H4S114, Harp, P311, PTZ17, SEZ17 |
Location: |
Chr.18:33437019-33464029 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrep | NM_001109988.1 | NP_001103458.1 | ||
NM_001109989.2 | NP_001103459.1 | |||
NM_001267717.1 | NP_001254646.1 | |||
NM_053078.4 | NP_444308.1 | |||
AK020629.1 | BAB32157.1 | |||
AK048361.1 | ||||
AK051015.1 | 1 | 1536 | ||
AK081502.1 | BAC38238.1 | |||
AK133769.1 | 1 | 1349 | BAE21830.1 | |
BC054762.1 | AAH54762.1 | |||
BQ715224.1 | ||||
BU057200.1 | ||||
CJ148521.1 | ||||
CJ165393.1 | ||||
CX768079.1 | ||||
EU447302.1 | ||||
X70398.1 | CAA49848.1 |
