Assay Details
Target Gene Details
Entrez Gene ID: | 77805 |
Gene Name: | establishment of cohesion 1 homolog 1 (S. cerevisiae) |
Gene Aliases: |
A930014I12Rik |
Location: |
Chr.18:10566507-10610685 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esco1 | NM_001081222.1 | NP_001074691.1 | ||
| AK040576.1 | ||||
| AK049589.1 | BAC33830.1 | |||
| AK050228.1 | 1 | 206 | ||
| AK139549.1 | ||||
| AK145333.1 | BAE26373.1 | |||
| AK145870.1 | BAE26712.1 | |||
| AK173297.1 | BAD32575.1 | |||
| BC008220.1 | AAH08220.1 | |||
| BC151069.1 | ||||
| BC151077.1 |
More Information
Set Membership: |
|

Genomic Map