Assay Details
Target Gene Details
Entrez Gene ID: | 211924 |
Gene Name: | desmoglein 1 gamma |
Gene Aliases: |
9330156N18, Dsg1gamma, Dsg6, dsg1g |
Location: |
Chr.18:20247340-20283923 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsg1c | NM_181680.1 | NP_859008.1 | ||
| AY192159.1 | AAP31153.1 |
More Information
Set Membership: |
|

Genomic Map