Assay Details
Target Gene Details
Entrez Gene ID: | 140492 |
Gene Name: | potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2 |
Gene Aliases: |
KCa2.2, SK2, SKCA2, fri |
Location: |
Chr.18:45268764-45685887 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnn2 | NM_080465.2 | NP_536713.1 | ||
| AF357240.1 | AAK48901.1 | |||
| AF533008.1 | AAQ10283.1 | |||
| AK033158.1 | BAC28176.1 | |||
| AK039033.1 | BAC30212.1 | |||
| AK050390.1 | BAC34231.1 | |||
| AK149082.1 | BAE28732.1 | |||
| AK215596.1 | ||||
| AK216215.1 | ||||
| AY123778.1 | AAM88568.1 | |||
| AY280964.1 | AAP31202.1 | |||
| BC125475.1 | ||||
| BC137656.1 | ||||
| EU478850.1 |
More Information
Set Membership: |
|

Genomic Map