Assay Details
Target Gene Details
Entrez Gene ID: | 75533 |
Gene Name: | NME/NM23 family member 5 |
Gene Aliases: |
1700019D05Rik, Nm23-M5 |
Location: |
Chr.18:34562636-34579152 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme5 | NM_080637.3 | 2 | 135 | NP_542368.2 |
| AF343565.1 | AAK20866.1 | |||
| AK006111.1 | 1 | 119 | ||
| AK131722.1 | 2 | 136 | BAE20779.1 | |
| BC049625.1 | 2 | 129 | AAH49625.1 |
More Information
Set Membership: |
|

Genomic Map