Assay Details
Target Gene Details
Entrez Gene ID: | 20522 |
Gene Name: | solute carrier family 23 (nucleobase transporters), member 1 |
Gene Aliases: |
D18Ucla2, SVCT1, Slc23a2, YSPL3 |
Location: |
Chr.18:35604041-35630340 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc23a1 | NM_011397.4 | 6 | 620 | NP_035527.3 |
| AF058318.1 | 6 | 530 | AAC78805.1 | |
| AK085499.1 | 6 | 535 | BAC39457.1 | |
| AK143959.1 | 6 | 621 | BAE25631.1 | |
| AK165439.1 | 6 | 556 | BAE38187.1 | |
| BC013528.1 | 6 | 549 | AAH13528.1 |
More Information
Set Membership: |
|

Genomic Map