Assay Details
Target Gene Details
Entrez Gene ID: | 319211 |
Gene Name: | nucleolar protein 4 |
Gene Aliases: |
1700013J13Rik, 4930568N03Rik, Gm1262 |
Location: |
Chr.18:22693152-23041675 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol4 | NM_001161483.1 | 4 | 433 | NP_001154955.1 |
| NM_199024.2 | 4 | 2203 | NP_950189.2 | |
| AK031008.1 | 4 | 2203 | ||
| BC056377.1 | 4 | 426 | AAH56377.1 |

Genomic Map