Assay Details
Target Gene Details
Entrez Gene ID: | 70640 |
Gene Name: | DCP2 decapping enzyme homolog (S. cerevisiae) |
Gene Aliases: |
2410015D23Rik, 5730537H01Rik, AL118268 |
Location: |
Chr.18:44380500-44424969 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dcp2 | NM_027490.1 | 9 | 1132 | NP_081766.1 |
AK017809.1 | 9 | 1124 | BAB30946.1 | |
AK152160.1 | 9 | 1105 | BAE30995.1 | |
AK153105.1 | 9 | 1134 | ||
AK153264.1 | 9 | 1105 | BAE31854.1 | |
AK153310.1 | 9 | 1105 | BAE31891.1 | |
AK166560.1 | 9 | 1111 | BAE38853.1 |
