Assay Details
Target Gene Details
Entrez Gene ID: | 15200 |
Gene Name: | heparin-binding EGF-like growth factor |
Gene Aliases: |
AW047313, Dtr, Dts, Hegfl |
Location: |
Chr.18:36504927-36515805 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hbegf | NM_010415.2 | 3 | 643 | NP_034545.1 |
| AK145548.1 | 3 | 612 | BAE26501.1 | |
| AK160575.1 | 3 | 613 | BAE35883.1 | |
| AK164309.1 | 3 | 644 | BAE37729.1 | |
| BC089607.1 | 3 | 612 | AAH89607.1 | |
| L07264.1 | 3 | 576 | AAA37542.1 |
More Information
Set Membership: |
|

Genomic Map