Assay Details
Target Gene Details
Entrez Gene ID: | 15183 |
Gene Name: | histone deacetylase 3 |
Gene Aliases: |
AW537363 |
Location: |
Chr.18:37936971-37955000 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac3 | NM_010411.2 | 13 | 1098 | NP_034541.2 |
AF074881.1 | 13 | 1045 | AAC36305.1 | |
AF074882.1 | 7 | 472 | AAC36306.1 | |
AF098295.1 | 13 | 1023 | AAF28798.1 | |
AF125536.1 | 13 | 1030 | AAF36425.1 | |
AK145158.1 | 13 | 1064 | BAE26265.1 | |
AK206656.1 | ||||
AK208847.1 | ||||
BC139300.1 | 13 | 1126 | ||
BC139301.1 | 13 | 1126 | ||
JN651901.1 | 7 | 746 | ||
JN651902.1 | 6 | 704 | ||
JN651903.1 | 7 | 455 | ||
JN651904.1 | 4 | 300 |
