Assay Details
Target Gene Details
Entrez Gene ID: | 15183 |
Gene Name: | histone deacetylase 3 |
Gene Aliases: |
AW537363 |
Location: |
Chr.18:37936971-37955000 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac3 | NM_010411.2 | NP_034541.2 | ||
AF074881.1 | AAC36305.1 | |||
AF074882.1 | AAC36306.1 | |||
AF098295.1 | AAF28798.1 | |||
AF125536.1 | AAF36425.1 | |||
AK145158.1 | BAE26265.1 | |||
BC139300.1 | ||||
BC139301.1 | ||||
BQ932476.1 | ||||
CJ166955.1 | ||||
JN651901.1 | ||||
JN651902.1 | ||||
JN651903.1 | ||||
JN651904.1 |
