Assay Details
Target Gene Details
Entrez Gene ID: | 18145 |
Gene Name: | Niemann-Pick type C1 |
Gene Aliases: |
A430089E03Rik, C85354, D18Ertd139e, D18Ertd723e, lcsd, nmf164, spm |
Location: |
Chr.18:12189694-12236386 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npc1 | NM_008720.2 | NP_032746.2 | ||
| AF003348.1 | AAB63372.1 | |||
| AK040367.1 | 2 | 283 | ||
| AK044878.1 | BAC32127.1 | |||
| AK087174.1 | ||||
| AK154282.1 | BAE32486.1 | |||
| AK155379.1 | BAE33230.1 | |||
| AK170880.1 | ||||
| AK171371.1 | 2 | 251 | ||
| BC052437.1 | AAH52437.1 | |||
| BC054539.1 | AAH54539.1 |

Genomic Map