Assay Details
Target Gene Details
Entrez Gene ID: | 269003 |
Gene Name: | Sin3A associated protein |
Gene Aliases: |
2610304F09Rik, 6720406D06 |
Location: |
Chr.18:31634327-31723068 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sap130 | NM_172965.2 | 4 | 537 | NP_766553.1 |
| AK034401.1 | 4 | 541 | ||
| AK053675.1 | 4 | 538 | BAC35469.1 | |
| AK084078.1 | 4 | 538 | BAC39113.1 | |
| BC063075.1 | AAH63075.1 | |||
| BC066030.1 | 4 | 549 | AAH66030.1 |
More Information
Set Membership: |
|

Genomic Map