Assay Details
Target Gene Details
Entrez Gene ID: | 240332 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter, L-proline), member 7 |
Gene Aliases: |
AW455934, PROT |
Location: |
Chr.18:60995376-61014199 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a7 | NM_201353.1 | NP_958741.1 | ||
| AK141581.1 | ||||
| BC050103.1 | AAH50103.1 | |||
| BC057070.1 | AAH57070.1 |
More Information
Set Membership: |
|

Genomic Map