Assay Details
Target Gene Details
Entrez Gene ID: | 170459 |
Gene Name: | StAR-related lipid transfer (START) domain containing 4 |
Gene Aliases: |
4632419C16Rik, 9030213J02Rik, AA517649, AW324468 |
Location: |
Chr.18:33199140-33213879 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stard4 | NM_133774.4 | NP_598535.1 | ||
| AF480297.1 | AAL87127.1 | |||
| AF480298.1 | AAL87128.1 | |||
| AK042347.1 | BAC31233.1 | |||
| AK042691.1 | BAC31335.1 | |||
| AK054501.1 | BAC35805.1 | |||
| AK078858.1 | BAC37423.1 | |||
| AK085331.1 | BAC39424.1 | |||
| AK165641.1 | ||||
| AK208141.1 | ||||
| BC005642.1 | AAH05642.1 |

Genomic Map