Assay Details
Target Gene Details
Entrez Gene ID: | 170461 |
Gene Name: | StAR-related lipid transfer (START) domain containing 6 |
Gene Aliases: |
1700011K09Rik, 4833424I06Rik, 4933429L05Rik, AI644424 |
Location: |
Chr.18:70472454-70505254 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stard6 | NM_001289648.1 | NP_001276577.1 | ||
NM_001289649.1 | NP_001276578.1 | |||
NM_001289650.1 | NP_001276579.1 | |||
NM_029019.4 | NP_083295.1 | |||
AK005873.1 | BAC25128.1 | |||
AK014759.1 | BAB29535.2 | |||
AK016984.1 | BAB30538.2 | |||
BC087893.1 | AAH87893.1 | |||
BY085273.1 |
