Assay Details
Target Gene Details
Entrez Gene ID: | 13527 |
Gene Name: | dystrobrevin alpha |
Gene Aliases: |
2210407P21Rik, DTN-A, Dtn, Gm19389, a-DB-1, adbn |
Location: |
Chr.18:23310020-23659719 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtna | NM_001285807.1 | 12 | 1614 | NP_001272736.1 |
| NM_001285808.1 | 10 | 1270 | NP_001272737.1 | |
| NM_001285810.1 | 11 | 1279 | NP_001272739.1 | |
| NM_001285811.1 | 11 | 1392 | NP_001272740.1 | |
| NM_001285813.1 | 12 | 1436 | NP_001272742.1 | |
| NM_001285817.1 | 10 | 1270 | NP_001272746.1 | |
| NM_010087.4 | 11 | 1392 | NP_034217.2 | |
| NM_207650.4 | 11 | 1392 | NP_997533.1 | |
| AF143542.1 | 11 | 1291 | AAD33913.1 | |
| AF143543.1 | 11 | 1328 | AAD33914.1 | |
| AF143544.1 | 11 | 1237 | AAD33915.1 | |
| AJ009669.1 | CAA08770.1 | |||
| AK003829.2 | 11 | 1312 | BAC25056.1 | |
| AK019477.1 | 12 | 1534 | BAB31746.1 | |
| AK131649.1 | 10 | 1163 | BAE20740.1 | |
| AK133036.1 | 2 | 115 | BAE21478.1 | |
| AK147279.1 | 12 | 1347 | BAE27816.1 | |
| AK147359.1 | 11 | 1236 | BAE27865.1 | |
| BC040364.1 | 11 | 1359 | AAH40364.1 | |
| BC050765.1 | 10 | 1178 | AAH50765.1 | |
| BU700818.1 | 2 | 323 | ||
| X95226.1 | 10 | 1092 | CAA64518.1 | |
| X95227.1 | 12 | 1231 | CAA64519.1 |
More Information
Set Membership: |
|

Genomic Map