Assay Details
Target Gene Details
Entrez Gene ID: | 70417 |
Gene Name: | multiple EGF-like-domains 10 |
Gene Aliases: |
3000002B06Rik, Gm331 |
Location: |
Chr.18:57133090-57297467 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 21 - Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Megf10 | NM_001001979.2 | NP_001001979.1 | ||
| AK147238.1 | BAE27788.1 | |||
| AK148084.1 | BAE28336.1 | |||
| AK166316.1 | BAE38699.1 | |||
| BC075647.1 | AAH75647.1 |
More Information
Set Membership: |
|

Genomic Map