Assay Details
Target Gene Details
| Entrez Gene ID: | 67869 | 
| Gene Name: | polyadenylate-binding protein-interacting protein 2 | 
| Gene Aliases: | 2310050K10Rik, AU045972 | 
| Location: | Chr.18:35598602-35617185 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Paip2 | NM_026420.2 | 2 | 178 | NP_080696.1 | 
| AK005462.1 | 2 | 180 | BAB24053.1 | |
| AK009909.1 | 2 | 179 | BAB26578.1 | |
| AK146327.1 | 2 | 209 | BAE27082.1 | |
| AK150957.1 | 2 | 186 | BAE29989.1 | |
| AK154453.1 | 3 | 280 | BAE32597.1 | |
| AK167545.1 | 2 | 205 | BAE39611.1 | |
| AK169207.1 | 2 | 200 | BAE40979.1 | |
| AK169411.1 | 2 | 200 | BAE41157.1 | |
| BC010190.1 | 2 | 194 | AAH10190.1 | 
Target Gene Details
| Entrez Gene ID: | 20522 | 
| Gene Name: | solute carrier family 23 (nucleobase transporters), member 1 | 
| Gene Aliases: | D18Ucla2, SVCT1, Slc23a2, YSPL3 | 
| Location: | Chr.18:35604041-35630340 on Build GRCm38 | 
| Assay Gene Location: | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Slc23a1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic