Assay Details
Target Gene Details
Entrez Gene ID: | 75599 |
Gene Name: | protocadherin 1 |
Gene Aliases: |
2010005A06Rik, AI585920 |
Location: |
Chr.18:38185914-38212048 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdh1 | NM_029357.3 | 1 | 101 | NP_083633.2 |
| AY861431.1 | AAX56121.1 | |||
| BC040402.1 | 1 | 101 | AAH40402.1 |
More Information
Set Membership: |
|

Genomic Map