Assay Details
Target Gene Details
Entrez Gene ID: | 20358 |
Gene Name: | sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A |
Gene Aliases: |
9330158E07, A730020P05Rik, AI851735, Sema6A-1, Semaq, VIa, b2b997Clo |
Location: |
Chr.18:47245254-47371487 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema6a | NM_018744.2 | NP_061214.2 | ||
| AF030430.1 | AAB86408.1 | |||
| AF288666.1 | AAG29494.1 | |||
| AK042751.1 | BAC31351.1 | |||
| AK082711.1 | BAC38582.1 | |||
| BC059238.1 | AAH59238.1 | |||
| BC062979.1 | AAH62979.1 |
More Information
Set Membership: |
|

Genomic Map