Assay Details
Target Gene Details
Entrez Gene ID: | 269016 |
Gene Name: | SH3 domain containing ring finger 2 |
Gene Aliases: |
2310046K19, 9130023G24Rik, AI452083, Ppp1r39, RNF158 |
Location: |
Chr.18:42051743-42158695 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh3rf2 | NM_001146299.1 | 7 | 1640 | NP_001139771.1 |
NM_172966.3 | 6 | 1544 | NP_766554.2 | |
AK028153.1 | 6 | 1040 | BAC25780.1 | |
AK029168.1 | 6 | 1545 | BAC26334.1 | |
AK033601.1 | 7 | 1684 | BAC28382.1 | |
BC048400.1 | 1 | 209 | AAH48400.1 | |
BC137955.1 | 7 | 1593 | ||
BC145060.1 | 7 | 1593 |
