Assay Details
Target Gene Details
| Entrez Gene ID: | 26379 | 
| Gene Name: | estrogen related receptor, alpha | 
| Gene Aliases: | ERRalpha, Err1, Estrra, Nr3b1 | 
| Location: | Chr.19:6910977-6922048 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Intron 3 - Exon 4 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Esrra | NM_007953.2 | NP_031979.2 | ||
| AK019222.1 | ||||
| AK156371.1 | BAE33690.1 | |||
| BC138586.1 | ||||
| BC171958.1 | ||||
| CO425338.1 | ||||
| U85259.1 | AAB51250.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic