Assay Details
Target Gene Details
Entrez Gene ID: | 76539 |
Gene Name: | family with sequence similarity 204, member A |
Gene Aliases: |
2310065H12Rik, 2610015K05Rik, D19Ertd737e |
Location: |
Chr.19:60198586-60226726 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam204a | NM_029648.6 | NP_083924.2 | ||
| AK011404.2 | BAB27596.2 | |||
| AK075914.1 | BAC36050.1 | |||
| AK162079.1 |

Genomic Map