Assay Details
Target Gene Details
Entrez Gene ID: | 66727 |
Gene Name: | HRAS-like suppressor family, member 5 |
Gene Aliases: |
1700055A14Rik, 1700086E08Rik, 4921526K24Rik, HRSL5, iNAT |
Location: |
Chr.19:7612569-7639238 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hrasls5 | NM_025731.2 | NP_080007.1 | ||
AB261168.1 | ||||
AB447517.1 | ||||
AK006527.1 | BAB24636.2 | |||
AK006790.1 | BAB24741.1 | |||
AK014971.1 | BAB29646.1 | |||
BC061008.1 | AAH61008.1 |
